MilliporeSigma
  • Alphav-integrin utilization in human beta-cell adhesion, spreading, and motility.
Alphav-integrin utilization in human beta-cell adhesion, spreading, and motility.
The Journal of biological chemistry (2004-02-10)
Thomas Kaido, Brandon Perez, Mayra Yebra, Jesse Hill, Vincenzo Cirulli, Alberto Hayek, Anthony M Montgomery
ABSTRACT
The role of individual integrins in human beta-cell development and function is largely unknown. This study describes the contribution of alpha(v)-integrins to human beta-cell adhesion, spreading, and motility. Developmental differences in alpha(v)-integrin utilization are addressed by comparing the responses of adult and fetal beta-cells, and vitronectin is used as a substrate based on its unique pattern of expression in the developing pancreas. Fetal and adult beta-cells attached equally to vitronectin and integrin alpha(v)beta(5) was found to support the adhesion of both mature and immature beta-cell populations. Fetal beta-cells were also observed to spread and migrate on vitronectin, and integrin alpha(v)beta(1) was found to be essential for these responses. In contrast to their fetal counterparts, adult beta-cells failed to either spread or migrate and this deficit was associated with a marked down-regulation of alpha(v)beta(1) expression in adult islet preparations. The integrin alpha(v)beta(3) was not found to support significant beta-cell attachment or migration. Based on our findings, we conclude that integrins alpha(v)beta(5) and alpha(v)beta(1) are important mediators of human beta-cell adhesion and motility, respectively. By supporting fetal beta-cell migration, alpha(v)beta(1) could play an important role in early motile processes required for islet neogenesis.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, FITC conjugated, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4C10, culture supernatant, clone P4C10, Chemicon®
Sigma-Aldrich
Anti-Integrin alphaV Antibody, CT, Intracellular, serum, Chemicon®