MilliporeSigma
  • The extracellular domain of CD11d regulates its cell surface expression.
The extracellular domain of CD11d regulates its cell surface expression.
Journal of leukocyte biology (2009-07-03)
William M McKillop, John W Barrett, Stephen H Pasternak, Bosco M C Chan, Gregory A Dekaban
ABSTRACT
A mAb targeting the CD11d subunit of the leukocyte integrin CD11d/CD18 decreases intraspinal inflammation and oxidative damage leading to improved neurological outcomes in rodent models of SCI. CD11d/CD18 is the fourth member of the beta2-integrin family. Current evidence indicates that CD11d/CD18 is regulated differently than other beta2-integrins, suggesting that CD11d(+) leukocytes play a distinct role in inflammation. Although the transcriptional control of CD11d expression has been evaluated, control of the intracellular distribution of CD11d has not been addressed. For this reason and as a result of the potential of CD11d as a therapeutic target for SCI and possibly other CNS injuries, we investigated the intracellular localization and surface expression of CD11d in cultured cells. CD11d and CD18 were fused at their C-termini with YFP and mRFP, respectively. Flow cytometry and confocal microscopy demonstrated that rCD11d-YFP is expressed on the cell surface of leukocyte cell lines expressing CD18. In contrast, in heterologous cell lines, CD11d-YFP is retained intracellularly in the TGN. Coexpression of CD11d-YFP and CD18-mRFP relieves this intracellular restriction and allows the CD11d/CD18 heterodimer to be surface-expressed. Based on domain-swapping experiments with CD25, the extracellular domain of CD11d is required and sufficient for the observed intracellular retention in heterologous cells. Furthermore, the transmembrane and C-terminus are also required for proper heterodimerization with CD18 and localization to the plasma membrane. These findings suggest that multiple CD11d domains play a role in controlling intracellular location and association with CD18.
Sigma-Aldrich
Anti-Integrin β2 [CD18] Antibody, clone MEM 48, Chemicon®, from mouse