MilliporeSigma
  • Site-specific nitration and oxidative dityrosine bridging of the tau protein by peroxynitrite: implications for Alzheimer's disease.
Site-specific nitration and oxidative dityrosine bridging of the tau protein by peroxynitrite: implications for Alzheimer's disease.
Biochemistry (2005-02-03)
Matthew R Reynolds, Robert W Berry, Lester I Binder
ABSTRACT
Alzheimer's disease (AD) is a progressive amnestic disorder typified by the pathological misfolding and deposition of the microtubule-associated tau protein into neurofibrillary tangles (NFTs). While numerous post-translational modifications influence NFT formation, the molecular mechanisms responsible for tau aggregation remain enigmatic. Since nitrative and oxidative injury have previously been shown to play a mechanistic role in neurodegeneration, we examined whether these events influence tau aggregation. In this report, we characterize the effects of peroxynitrite (ONOO-)-mediated nitration and oxidation on tau polymerization in vitro. Treatment of tau with ONOO- results in 3-nitrotyrosine (3-NT) immunoreactivity and the formation of heat-stable, SDS-insoluble oligomers. Using ESI-MS and HPLC with fluorescent detection, we show that these higher-order aggregates contain 3,3'-dityrosine (3,3'-DT). Tyrosine (Tyr) residues are critical for ONOO(-)-mediated oligomerization, as tau proteins lacking all Tyr residues fail to generate oligomers upon ONOO- treatment. Further, tau nitration targets residues Y18, Y29, and to a lesser degree Y197 and Y394, and nitration at these sites inhibits in vitro polymerization. The inhibitory effect of nitration on tau polymerization is specific for the 3-NT modification, as pseudophosphorylation at these same Tyr residues does not inhibit tau assembly. Our results suggest that the nitrative and oxidative roles of ONOO- differentially affect tau polymerization and that ONOO(-)-mediated cross-linking could facilitate tau aggregation in AD.
Sigma-Aldrich
Thrombin CleanCleave Kit