MilliporeSigma
Animal models of neurodegenerative diseases.
Methods in molecular biology (Clifton, N.J.) (2009-04-21)
Wendy Phillips, Andrew Michell, Harald Pruess, Roger A Barker
ABSTRACT
Animal models of neurodegenerative disease are excellent tools for studying pathogenesis and therapies including cellular transplantation. In this chapter, we describe different models of Huntington's disease and Parkinson's disease, stereotactic surgery (used in creation of lesion models and transplantation) and finally transplantation studies in these models.
Sigma-Aldrich
β-Galactosidase Reporter Gene Staining Kit
Pricing and availability is not currently available.