MilliporeSigma
  • Radiosensitization of MDA-MB-231 breast tumor cells by adenovirus-mediated overexpression of a fragment of the XRCC4 protein.
Radiosensitization of MDA-MB-231 breast tumor cells by adenovirus-mediated overexpression of a fragment of the XRCC4 protein.
Molecular cancer therapeutics (2005-10-18)
Kara R Jones, David A Gewirtz, Steven M Yannone, Shaoming Zhou, David G Schatz, Kristoffer Valerie, Lawrence F Povirk
ABSTRACT
Incomplete DNA repair or misrepair can contribute to the cytotoxicity of DNA double-strand breaks. Consequently, interference with double-strand break repair, by pharmacologic or genetic means, is likely to sensitize tumor cells to ionizing radiation. The current studies were designed to inhibit the nonhomologous end joining repair pathway by interfering with the function of the XRCC4/ligase IV complex. A PCR-generated fragment of the XRCC4 gene, encompassing the homodimerization and ligase IV-binding domains, was inserted into a plasmid vector (pFLAG-CMV-2) expressing the FLAG peptide and the cassette encoding FLAG-tagged XRCC4 fragment was cloned into an adenoviral vector. Both the plasmid and the corresponding adenovirus elicited robust expression of a truncated XRCC4 protein designed to compete in a dominant-negative fashion with full-length XRCC4 for binding to ligase IV. Binding of the XRCC4 fragment to ligase IV in vivo was confirmed by immunoprecipitation. Clonogenic survival assays showed that the adenovirus expressing the truncated XRCC4 protein sensitizes MDA-MB-231 breast tumor cells to ionizing radiation, presumably through interference with the functional activity of ligase IV, leading to inhibition of the final ligation step in end joining. These studies support the potential clinical utility of combining radiation therapy with agents that inhibit DNA double-strand break repair.
Product Description
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)