MilliporeSigma
  • The dopamine D2 receptor: two molecular forms generated by alternative splicing.
The dopamine D2 receptor: two molecular forms generated by alternative splicing.
The EMBO journal (1989-12-20)
R Dal Toso, B Sommer, M Ewert, A Herb, D B Pritchett, A Bach, B D Shivers, P H Seeburg
ABSTRACT
Cloned human dopamine D2 receptor cDNA was isolated from a pituitary cDNA library and found to encode an additional 29 amino acid residues in the predicted intracellular domain between transmembrane regions 5 and 6 relative to a previously described rat brain D2 receptor. Results from polymerase chain reactions as well as in situ hybridization revealed that mRNA encoding both receptor forms is present in pituitary and brain of both rat and man. The larger form was predominant in these tissues and, as shown in the rat, expressed by dopaminergic and dopaminoceptive neurons. Analysis of the human gene showed that the additional peptide sequence is encoded by a separate exon. Hence, the two receptor forms are generated by differential splicing possibly to permit coupling to different G proteins. Both receptors expressed in cultured mammalian cells bind [3H]spiperone with high affinity and inhibit adenylyl cyclase, as expected of the D2 receptor subtype.
Sigma-Aldrich
Dopamine D2 Receptor, control peptide for AB5084P