MilliporeSigma
  • Mild conversion of beta-diketones and beta-ketoesters to carboxylic acids.
Mild conversion of beta-diketones and beta-ketoesters to carboxylic acids.
The Journal of organic chemistry (2006-06-06)
Yang Zhang, Jingliang Jiao, Robert A Flowers
ABSTRACT
A mild protocol for the conversion of beta-ketoesters and beta-diketones to carboxylic acids with use of CAN in CH3CN is described. The method is compatible with a number of functional groups, and can generate carboxylic acids under neutral conditions at room temperature. The reaction is fast and general, providing an alternative method to the commonly used malonic ester acid preparation. Initial mechanistic studies show that initial oxidation of the enol form of the beta-dicarbonyl initiates the reaction. The presence of nitrate as an oxidant ligand or as an additive is critical for success of the reaction.
Sigma-Aldrich
3-Methoxybenzyl bromide, 98%