MilliporeSigma
  • Prmt5 is required for germ cell survival during spermatogenesis in mice.
Prmt5 is required for germ cell survival during spermatogenesis in mice.
Scientific reports (2015-06-16)
Yanbo Wang, Tianxiang Zhu, Qiuling Li, Chunyi Liu, Feng Han, Min Chen, Lianjun Zhang, Xiuhong Cui, Yan Qin, Shilai Bao, Fei Gao
ABSTRACT
During germ cell development, epigenetic modifications undergo extensive remodeling. Abnormal epigenetic modifications usually result in germ cell loss and reproductive defect. Prmt5 (Protein arginine methyltransferase 5) encodes a protein arginine methyltransferase which has been demonstrated to play important roles in germ cell development during embryonic stages. In the present study, we found that Prmt5 was also abundantly expressed in male germ cells after birth. Inactivation of this gene by crossing with Stra8-Cre transgenic mice resulted in germ cell loss during spermatogenesis. Further study revealed that the germ cell development was grossly normal before P10. However, most of the germ cells in Prmt5(Δ/f); Stra8-Cre mice were blocked at meiotic stage. The expression of meiosis associated genes was reduced in Prmt5(Δ/f); Stra8-Cre testes compared to control testes at P10. γH2AX was detected in sex body of control germ cells at P12, whereas multiple foci were observed in Prmt5-deficient germ cells. Further study revealed that H4R3me2s was virtually absent in germ cells after Prmt5 inactivation. The results of this study indicate that Prmt5 also plays important roles in germ cell development during spermatogenesis.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-PRMT5 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl Histone H3 (Arg2), symmetric Antibody, serum, from rabbit