MilliporeSigma
  • One-pot synthesis of new substituted 1,2,3,4-tetrahydrocarbazoles via Petasis reaction.
One-pot synthesis of new substituted 1,2,3,4-tetrahydrocarbazoles via Petasis reaction.
Journal of combinatorial chemistry (2010-07-01)
Subhasish Neogi, Amrita Roy, Dinabandhu Naskar
ABSTRACT
The one-pot synthesis of a new substituted 1,2,3,4-tetrahydrocarbazoles has been described via Petasis reactions. These tetrahydrocarbazoles exhibits various medicinal importance and might be suitable for elaboration into larger peptides at carboxy termini. The scope and limitations of this method have been examined.