MilliporeSigma
  • Sensory-related neural activity regulates the structure of vascular networks in the cerebral cortex.
Sensory-related neural activity regulates the structure of vascular networks in the cerebral cortex.
Neuron (2014-08-27)
Baptiste Lacoste, Cesar H Comin, Ayal Ben-Zvi, Pascal S Kaeser, Xiaoyin Xu, Luciano da F Costa, Chenghua Gu
ABSTRACT
Neurovascular interactions are essential for proper brain function. While the effect of neural activity on cerebral blood flow has been extensively studied, whether or not neural activity influences vascular patterning remains elusive. Here, we demonstrate that neural activity promotes the formation of vascular networks in the early postnatal mouse barrel cortex. Using a combination of genetics, imaging, and computational tools to allow simultaneous analysis of neuronal and vascular components, we found that vascular density and branching were decreased in the barrel cortex when sensory input was reduced by either a complete deafferentation, a genetic impairment of neurotransmitter release at thalamocortical synapses, or a selective reduction of sensory-related neural activity by whisker plucking. In contrast, enhancement of neural activity by whisker stimulation led to an increase in vascular density and branching. The finding that neural activity is necessary and sufficient to trigger alterations of vascular networks reveals an important feature of neurovascular interactions.
Sigma-Aldrich
Anti-NeuN Antibody, serum, from guinea pig