MilliporeSigma
  • Room temperature palladium-catalyzed cross coupling of aryltrimethylammonium triflates with aryl Grignard reagents.
Room temperature palladium-catalyzed cross coupling of aryltrimethylammonium triflates with aryl Grignard reagents.
Organic letters (2010-09-04)
Jonathan T Reeves, Daniel R Fandrick, Zhulin Tan, Jinhua J Song, Heewon Lee, Nathan K Yee, Chris H Senanayake
ABSTRACT
Aryltrimethylammonium triflates and tetrafluoroborates were found to be highly reactive electrophiles in the Pd-catalyzed cross coupling with aryl Grignard reagents. The coupling reactions proceed at ambient temperature with a nearly stoichiometric quantity of Grignard reagent, and diverse functionality is tolerated. Competition experiments established the reactivity of PhNMe(3)OTf relative to PhCl, PhBr, PhI, and PhOTf.
Sigma-Aldrich
Methyl trifluoromethanesulfonate, ≥98%