MilliporeSigma
  • Discovery of boronic acids as novel and potent inhibitors of fatty acid amide hydrolase.
Discovery of boronic acids as novel and potent inhibitors of fatty acid amide hydrolase.
Journal of medicinal chemistry (2008-11-06)
Anna Minkkilä, Susanna M Saario, Heikki Käsnänen, Jukka Leppänen, Antti Poso, Tapio Nevalainen
ABSTRACT
A series of commercial phenyl-, heteroaryl-, alkyl-, and alkenylboronic acids were evaluated for their FAAH and MGL inhibitory activities. The compounds were generally selective for FAAH, with IC50 in the nanomolar or low-micromolar range. Eight of these compounds inhibited MGL with IC50 in the micromolar range. The most potent compound, phenylboronic acid with para-nonyl substituent (13), inhibited FAAH and MGL with IC50 of 0.0091 and 7.9 microM, respectively.
Sigma-Aldrich
Phenylboronic acid, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzo[b]thien-2-ylboronic acid, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
3-(Trifluoromethyl)phenylboronic acid, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Biphenylboronic acid, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Methoxyphenylboronic acid, ≥95.0%
Pricing and availability is not currently available.