MilliporeSigma
  • Fenoldopam preconditioning: role of heme oxygenase-1 in protecting human tubular cells and rodent kidneys against cold-hypoxic injury.
Fenoldopam preconditioning: role of heme oxygenase-1 in protecting human tubular cells and rodent kidneys against cold-hypoxic injury.
Transplantation (2010-11-16)
Abdulla K Salahudeen, Ming Yang, Hong Huang, Sylvain Dore, David E Stec
ABSTRACT
Kidneys from brain-dead donors are cold preserved until transplanted. However, prolonged cold storage can contribute to allograft failure. Studies suggest that donor preconditioning with dopaminergics may reduce cold-ischemic transplant injury, but whether heme oxygenase (HO)-1 induction is an underlying mechanism is not known. To test whether preconditioning with fenoldopam (FD) induce HO-1 and protect kidneys against cold storage injury and whether HO-1 plays a role in protection. We used human renal proximal tubular epithelial cells, rat kidney transplants, and HO-1 null mice kidneys. FD preconditioning of cells for 4 hr significantly protected against cell death from 24-hr cold hypoxia and was associated with a dose-dependent increase in HO-1 expression. In a syngeneic rat kidney transplant model, FD preconditioning for 18 hr markedly increased kidney HO-1 expression and protected kidneys against 24-hr cold-ischemic transplant injury. To test the role of HO-1, renal proximal tubular epithelial cells were treated with HO-1 small interfering RNA, followed by FD-preconditioning. Small interfering RNA inhibited the HO-1 messenger RNA expression and reversed the FD protection. Suspension of kidneys of HO-1 null and wild-type mice preconditioned with FD or saline were subjected to 24- and 48-hr cold storage. N-acetyl glucosaminidase, a specific tubular injury marker, was significantly lower in FD-preconditioned wild-type kidneys, but not in HO-1 null kidneys, suggesting a role for HO-1 in FD's preconditioning. Our data suggest HO-1 induction as an underlying mechanism for FD preconditioning and support the idea of testing FD preconditioning in the clinical setting. Studies are required to determine the optimum FD-preconditioning protocol.
Sigma-Aldrich
Anti-Heme Oxygenase-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture