MilliporeSigma
  • The liberation of fractalkine in the dorsal horn requires microglial cathepsin S.
The liberation of fractalkine in the dorsal horn requires microglial cathepsin S.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2009-05-29)
Anna K Clark, Ping K Yip, Marzia Malcangio
ABSTRACT
Understanding of the sequence and nature of the events that govern neuron-microglia communication is critical for the discovery of new mechanisms and targets for chronic pain treatment. The neuronal chemokine fractalkine (FKN) and its microglial receptor CX3CR1 may mediate such a function in the dorsal horn of the spinal cord after cleavage of the extracellular domain of this transmembrane chemokine by a protease. Here we report that in neuropathic rat dorsal horn, with dorsal root-attached preparations, soluble FKN (sFKN) contents are increased in the superfusates collected after noxious-like electrical stimulation of ipsilateral primary afferent fibers. The increase of sFKN is prevented by morpholinurea-leucine-homophenylalanine-vinyl sulfone-phenyl (LHVS), an irreversible inhibitor of cathepsin S (CatS) whose proteolytic activity is also increased in the superfusates. The source of CatS activity is microglial cells activated by the peripheral nerve injury and secreting the enzyme, as a result of primary afferent fiber stimulation. Indeed, the acute activation of dorsal horn microglia by lipopolysaccharide results in increased CatS activity in the superfusates, followed by increased sFKN contents. Consistent with these observations ex vivo, the levels of both sFKN and CatS activity in CSF samples increased significantly after peripheral nerve injury, associated with spinal microglial activation. Finally, because we found that both FKN immunoreactivity and mRNA are confined to dorsal horn neurons, we suggest that under neuropathic conditions, noxious stimulation of primary afferent fibers induces release of CatS from microglia, which liberates FKN from dorsal horn neurons, thereby contributing to the amplification and maintenance of chronic pain.
Sigma-Aldrich
Anti-Neurofilament 200 kDa Antibody, clone RT97, clone RT97, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse