MilliporeSigma
Bis(8-anilinonaphthalene-1-sulfonate) as a probe for tubulin decay.
Biochemistry (1986-02-11)
A R Prasad, R F Luduena, P M Horowitz
ABSTRACT
The fluorescent apolar probe bis(8-anilinonaphthalene-1-sulfonate) (Bis-ANS) has been used to detect structural correlates of the well-known but poorly understood decay of tubulin function, by which tubulin loses its ability to polymerize and bind drugs in a complex, time-dependent way. The present results indicate that the decay of tubulin is accompanied by the appearance of hydrophobic areas, which bind a total of six Bis-ANS molecules with a dissociation constant of 19 microM. This binding seems to be a result of localized structural changes that are taking place in the tubulin molecule and can be used as a probe for these changes. In particular, circular dichroism measurements revealed no significant changes in the average secondary structure of the protein during the time required for complete binding of the Bis-ANS molecules. Preincubation of tubulin with the antimitotic drugs colchicine, podophyllotoxin, and vinblastine slows the rate of appearance of the hydrophobic region. Vinblastine has the maximal effect followed by colchicine and podophyllotoxin. In contrast, preincubation with maytansine has no effect. In addition, lowering the temperature decreases the rate of appearance of this region. These results correlate with the effect of drugs on the alkylation of tubulin sulfhydryl groups by iodoacetamide [Luduena, R.F., & Roach, M.C. (1981) Biochemistry 20, 4444-4450] and with the ability of inhibitors of microtubule assembly to permit the polymerization of tubulin into nonmicrotubule structures.