MilliporeSigma
  • Scope of the Suzuki-Miyaura cross-coupling reactions of potassium heteroaryltrifluoroborates.
Scope of the Suzuki-Miyaura cross-coupling reactions of potassium heteroaryltrifluoroborates.
The Journal of organic chemistry (2008-12-25)
Gary A Molander, Belgin Canturk, Lauren E Kennedy
ABSTRACT
A wide variety of bench-stable potassium heteroaryltrifluoroborates were prepared, and general reaction conditions were developed for their cross-coupling to aryl and heteroaryl halides. The cross-coupled products were obtained in good to excellent yields. This method represents an efficient and facile installation of heterocyclic building blocks onto preexisting organic substructures.
Sigma-Aldrich
Tetrafluoroboric acid solution, 48 wt. % in H2O