MilliporeSigma
  • Nickel-catalyzed cross-coupling of potassium aryl- and heteroaryltrifluoroborates with unactivated alkyl halides.
Nickel-catalyzed cross-coupling of potassium aryl- and heteroaryltrifluoroborates with unactivated alkyl halides.
Organic letters (2010-11-26)
Gary A Molander, O Andreea Argintaru, Ioana Aron, Spencer D Dreher
ABSTRACT
A method for the cross-coupling of alkyl electrophiles with various potassium aryl- and heteroaryltrifluoroborates has been developed. Nearly stoichiometric amounts of organoboron species could be employed to cross-couple a large variety of challenging heteroaryl nucleophiles. Several functional groups were tolerated on both the electrophilic and the nucleophilic partners. Chemoselective reactivity of C(sp(3))-Br bonds in the presence of C(sp(2))-Br bonds was achieved.
Sigma-Aldrich
2-(Bromomethyl)tetrahydro-2H-pyran, 98%