MilliporeSigma
  • The STAT3-independent signaling pathway by glycoprotein 130 in hepatic cells.
The STAT3-independent signaling pathway by glycoprotein 130 in hepatic cells.
The Journal of biological chemistry (1999-03-13)
C F Lai, J Ripperger, Y Wang, H Kim, R B Hawley, H Baumann
ABSTRACT
Interleukin (IL)-6 is a major regulator of hepatic acute-phase plasma protein (APP) genes. The membrane-proximal 133-amino acid cytoplasmic domain of glycoprotein (gp) 130, containing one copy of the Box3 motif, is sufficient to transmit a productive signal to endogenous APP genes in rat hepatoma H-35 cells. In contrast, a mutant gp130 domain lacking the Box3 motif activates Janus kinases to a normal level but fails to activate signal transducer and activator of transcription 3 and to up-regulate a number of APP genes, including thiostatin, fibrinogen, hemopexin, and haptoglobin. However, in the absence of Box3, gp130 still stimulates the expression of alpha2-macroglobulin and synergizes with IL-1 to up-regulate alpha1-acid glycoprotein. The Box3 motif is not required for activation of the SH2-containing protein tyrosine phosphatase 2 or the mitogen-activated protein kinase (MAPK), nor is the immediate induction of egr-1 and junB significantly altered. Surprisingly, gp130 without any functional Box3 stimulates prolonged activation of MAPK, leading to an extended period of up-regulation of egr-1 and to an extracellularly regulated kinase-mediated reduction in the IL-6-stimulated production of thiostatin. IL-6 reduces proliferation of H-35 cells through signaling by the Box3. In addition, cells expressing Box3-deficient gp130 showed distinct morphologic changes upon receptor activation. Taken together, these results indicate that Box3-derived and Box3-independent signals cooperate in the control of hepatic APP genes and that Box3 may be involved in the modulation of MAPK activity in gp130 signaling.