MilliporeSigma
  • A phosphatidylinositol 3-kinase-dependent pathway that differentially regulates c-Raf and A-Raf.
A phosphatidylinositol 3-kinase-dependent pathway that differentially regulates c-Raf and A-Raf.
The Journal of biological chemistry (1999-03-06)
S L Sutor, B T Vroman, E A Armstrong, R T Abraham, L M Karnitz
ABSTRACT
Cytokines trigger the rapid assembly of multimolecular signaling complexes that direct the activation of downstream protein kinase cascades. Two protein kinases that have been linked to growth factor-regulated proliferation and survival are mitogen-activated protein/ERK kinase (MEK) and its downstream target Erk, a member of the mitogen-activated protein kinase family. Using complementary pharmacological and genetic approaches, we demonstrate that MEK and Erk activation requires a phosphatidylinositol 3-kinase (PI3-K)-generated signal in an interleukin (IL)-3-dependent myeloid progenitor cell line. Analysis of the upstream pathway leading to MEK activation revealed that inhibition of PI3-K did not block c-Raf activation, whereas MEK activation was effectively blocked under these conditions. Furthermore, agents that elevated cAMP suppressed IL-3-induced c-Raf activation but did not inhibit MEK activation. Because c-Raf activation and MEK activation were inversely affected by PI3-K- and cAMP-dependent pathways, we examined whether IL-3 activated the alternative Raf isoforms A-Raf and B-Raf. Although IL-3 did not activate B-Raf, A-Raf was activated by the cytokine. Moreover, A-Raf activation, like MEK activation, was blocked by inhibition of PI3-K but was insensitive to cAMP. Experiments with dominant negative mutants of the Raf isoforms showed that overexpression of dominant negative c-Raf did not prevent MEK activation. However, dominant negative A-Raf effectively blocked MEK activation, suggesting that activation of the MEK-Erk signaling cascade is mediated through A-Raf. Taken together, these results suggest that IL-3 receptors engage and activate both c-Raf and A-Raf in hemopoietic cells. However, these intermediates are differentially regulated by upstream signaling cascades and selectively coupled to downstream signaling pathways.