MilliporeSigma
  • Targeting the prostaglandin F2α receptor for preventing preterm labor with azapeptide tocolytics.
Targeting the prostaglandin F2α receptor for preventing preterm labor with azapeptide tocolytics.
Journal of medicinal chemistry (2011-07-22)
Carine B Bourguet, Eugénie Goupil, Danaë Tassy, Xin Hou, Eryk Thouin, Felix Polyak, Terence E Hébert, Audrey Claing, Stéphane A Laporte, Sylvain Chemtob, William D Lubell
ABSTRACT
The prostaglandin-F2α (PGF2α) receptor (FP) was targeted to develop tocolytic agents for inhibiting preterm labor. Azabicycloalkane and azapeptide mimics 2-10 were synthesized based on the (3S,6S,9S)-indolizidin-2-one amino acid analogue PDC113.824 (1), which was shown to modulate FP by a biased allosteric mechanism, involving both Gαq- and Gα12-mediated signaling pathways, and exhibited significant tocolytic activity delaying preterm labor in a mouse model ( Goupil ; et al. J. Biol. Chem. 2010 , 285 , 25624 - 25636 ). Although changes in azabicycloalkane stereochemistry and ring size caused loss of activity, replacement of the indolizidin-2-one amino acid with azaGly-Pro and azaPhe-Pro gave azapeptides 6 and 8, which reduced PGF2α-induced myometrial contractions, potentiated the effect of PGF2α on Gαq-mediated ERK1/2 activation, and inhibited FP modulation of cell ruffling, a response dependent on the Gα12/RhoA/ROCK signaling pathway. Revealing complementarities of azabicycloalkane and azapeptide mimics, novel probes, and efficient tocolytic agents were made to study allosteric modulation of the FP receptor.
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis