MilliporeSigma
  • Rainbow trout leucocyte activity: influence on the ectoparasitic monogenean Gyrodactylus derjavini
Rainbow trout leucocyte activity: influence on the ectoparasitic monogenean Gyrodactylus derjavini
Diseases of Aquatic Organisms (1999)
Buchmann K and Bresciani J