MilliporeSigma
  • Lewis Acid Catalyzed Borotropic Shifts in the Design of Diastereo- and Enantioselective γ-Additions of Allylboron Moieties to Aldimines.
Lewis Acid Catalyzed Borotropic Shifts in the Design of Diastereo- and Enantioselective γ-Additions of Allylboron Moieties to Aldimines.
Angewandte Chemie (International ed. in English) (2016-03-11)
Farid W van der Mei, Hiroshi Miyamoto, Daniel L Silverio, Amir H Hoveyda
ABSTRACT
Catalytic allylboron additions to aldimines are presented for which small amounts of Zn(OMe)2 serve as the co-catalyst to accelerate allyl exchange and 1,3-borotropic shift processes. Low-yielding and moderately α- and diastereoselective reactions are thus turned into highly efficient γ-, diastereo-, and enantioselective transformations that exhibit considerable scope.