MilliporeSigma
Hybrid Indicators for Fast and Sensitive Voltage Imaging.
Angewandte Chemie (International ed. in English) (2018-02-14)
Yongxian Xu, Luxin Peng, Sicong Wang, Anqi Wang, Ruirui Ma, Ying Zhou, Jiahe Yang, De-En Sun, Wei Lin, Xing Chen, Peng Zou
ABSTRACT
Membrane voltage is an important biophysical signal that underlies intercellular electrical communications. A fluorescent voltage indicator is presented that enables the investigation of electrical signaling at high spatial resolution. The method is built upon the site-specific modification of microbial rhodopsin proteins with organic fluorophores, resulting in a hybrid indicator scaffold that is one of the most sensitive and fastest orange-colored voltage indicators developed to date. We applied this technique to optically map electrical connectivity in cultured cells, which revealed gap junction-mediated long-range coupling that spanned over hundreds of micrometers.