MilliporeSigma
CDK5 Functions as a Tumor Promoter in Human Lung Cancer.
Journal of Cancer (2018-11-10)
Jie Zeng, Shuanshuan Xie, Yang Liu, Changxing Shen, Xiaolian Song, Guo-Lei Zhou, Changhui Wang
ABSTRACT
Cyclin-dependent kinase 5 (CDK5), an atypical member of the cyclin-dependent kinase family, plays an important role in the nervous system. Recent studies have shown that CDK5 is also associated with tumors. However, few studies have been done to investigate the mechanism underlying the connection between CDK5 and cancers. To explore the role of CDK5 in cancers by using an extensive bioinformatics data mining process. We mined the transcriptional, survival, functions and structure of CDK5 gene through databases and in vitro experiments. We found that higher CDK5 expression levels in most cancer cell lines while lower expression in liver and brain cancer cell lines. High expression of CDK5 was associated with shorter overall survival (OS) in lung cancer. In addition, high expression level of CDK5 promoted lung cancer cells proliferation and metastasis. Inhibited CDK5 decreases CAP1 phosphorylation. CDK5 may prove to be a valid target of anticancer therapies.
Sigma-Aldrich
MISSION® esiRNA, targeting human CDK5