MilliporeSigma
  • Blood-brainbarrier disruption dictates nanoparticle accumulation following experimental brain injury.
Blood-brainbarrier disruption dictates nanoparticle accumulation following experimental brain injury.
Nanomedicine : nanotechnology, biology, and medicine (2018-06-23)
Vimala N Bharadwaj, Rachel K Rowe, Jordan Harrison, Chen Wu, Trent R Anderson, Jonathan Lifshitz, P David Adelson, Vikram D Kodibagkar, Sarah E Stabenfeldt
ABSTRACT
Clinically, traumatic brain injury (TBI) results in complex heterogeneous pathology that cannot be recapitulated in single pre-clinical animal model. Therefore, we focused on evaluating utility of nanoparticle (NP)-based therapeutics following three diffuse-TBI models: mildclosed-head injury (mCHI), repetitive-mCHI and midline-fluid percussion injury (FPI). We hypothesized that NP accumulation after diffuse TBI correlates directly with blood-brainbarrier permeability. Mice received PEGylated-NP cocktail (20-500 nm) (intravenously) after single- or repetitive-(1 impact/day, 5 consecutive days) CHI (immediately) and midline-FPI (1 h, 3 h and 6 h). NPs circulated for 1 h before perfusion/brain extraction. NP accumulation was analyzed using fluorescent microscopy in brain regions vulnerable to neuropathology. Minimal/no NP accumulation after mCHI/RmCHI was observed. In contrast, midlineFPI resulted in significant peak accumulation of up to 500 nm NP at 3 h post-injury compared to sham, 1 h, and 6 h groups in the cortex. Therefore, our study provides the groundwork for feasibility of NP-delivery based on NPinjection time and NPsize after mCHI/RmCHI and midline-FPI.
Sigma-Aldrich
Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Methoxypolyethylene glycol amine, 20,000
Sigma-Aldrich
Methoxypolyethylene glycol amine, 2,000, extent of labeling: ≥0.4 mmol/g NH2 loading
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
MES hemisodium salt, ≥98% (titration)