MilliporeSigma
  • Combined Effect of Silver Nanoparticles, Ceftriaxone, and Methyluracil during Experimental Purulent Peritonitis.
Combined Effect of Silver Nanoparticles, Ceftriaxone, and Methyluracil during Experimental Purulent Peritonitis.
Bulletin of experimental biology and medicine (2018-10-26)
T V Chestnova, V G Zilov, P G Gladkikh, A A Khadartsev, A S Korotkova, Yu P Buzulukov
ABSTRACT
Bacterial biofilms provoke and/or promote the most chronic and recurrent infectious diseases. Previously, experimental models of purulent peritonitis and meningoencephalitis revealed positive antibiofilm effect of metallic nanoparticles and the absence of resistance against such nanoparticles in microorganisms. This study examines the combined effect of silver nanoparticles with ceftriaxone and methyluracil on recovery mechanisms during inflammatory diseases exemplified by purulent peritonitis in experimental animals.
Sigma-Aldrich
3-Methyluracil