MilliporeSigma
  • SODB1 is essential for Leishmania major infection of macrophages and pathogenesis in mice.
SODB1 is essential for Leishmania major infection of macrophages and pathogenesis in mice.
PLoS neglected tropical diseases (2018-10-30)
Bennett J Davenport, Casey G Martin, Stephen M Beverley, David J Orlicky, Andres Vazquez-Torres, Thomas E Morrison
ABSTRACT
Leishmania species are sand fly-transmitted protozoan parasites that cause leishmaniasis, neglected tropical diseases that affect millions of people. Leishmania amastigotes must overcome a variety of host defenses, including reactive oxygen species (ROS) produced by the NADPH oxidase. Leishmania species encode three superoxide dismutases (SODs): the mitochondrial SODA and two glycosomal SODs (SODB1 and SODB2). SODs are metalloenzymes that function in antioxidant defense by converting superoxide to oxygen and hydrogen peroxide. Here, we investigated a role for SODB1 in Leishmania infection of macrophages and virulence in mice. We found that a single allele deletion of SODB1 (SODB1/Δsodb1) had minimal effects on the replication of axenically-grown L. major promastigotes or differentiation to infective metacyclic promastigotes. Disruption of a single SODB1 allele also did not affect L. donovani differentiation to amastigotes induced axenically, or the replication of axenically-grown L. donovani promastigotes and amastigotes. In contrast, the persistence of SODB1/Δsodb1 L. major in WT macrophages was impaired, and the development of cutaneous lesions in SODB1/Δsodb1 L. major-infected C57BL/6 and BALB/c mice was strongly reduced. The reduced disease severity in mice was associated with reduced burdens of SODB1/Δsodb1 L. major parasites in the foot at late, but not early times post-inoculation, as well as an impaired capacity to disseminate from the site of inoculation. Collectively, these data suggest that SODB1 is critical for L. major persistence in macrophages and virulence in mice.
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Phleomycin from Streptomyces verticillus, powder
Sigma-Aldrich
Wright-Giemsa Stain, Modified
Sigma-Aldrich
RPMI 1640 Amino Acids Solution (50×), Without L-glutamine, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
RPMI 1640 Vitamins Solution (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, powder, BioReagent, suitable for cell culture, suitable for insect cell culture