MilliporeSigma
  • The pathway of transmembrane cadmium influx via calcium-permeable channels and its spatial characteristics along rice root.
The pathway of transmembrane cadmium influx via calcium-permeable channels and its spatial characteristics along rice root.
Journal of experimental botany (2018-08-14)
Xiaohui Chen, Younan Ouyang, Yicong Fan, Boyin Qiu, Guoping Zhang, Fanrong Zeng
ABSTRACT
To develop elite crops with low cadmium (Cd), a fundamental understanding of the mechanism of Cd uptake by crop roots is necessary. Here, a new mechanism for Cd2+ entry into rice root cells was investigated. The results showed that Cd2+ influx in rice roots exhibited spatially and temporally dynamic patterns. There was a clear longitudinal variation in Cd uptake along rice roots, with the root tip showing much higher Cd2+ influx and concentration than the root mature zone, which might be due to the much higher expression of the well-known Cd transporter genes OsIRT1, OsNRAMP1, OsNRAMP5, and OsZIP1 in the root tip. Both the net Cd2+ influx and the uptake of Cd in rice roots were highly inhibited by ion channel blockers Gd3+ and TEA+, supplementation of Ca2+ and K+, and the plasma membrane H+-ATPase inhibitor vanadate, with Gd3+ and Ca2+ showing the most inhibitory effects. Furthermore, Ca2+- or Gd3+-induced reduction in Cd2+ influx and Cd uptake did not coincide with the expression of Cd transporter genes, but with that of two Ca channel genes, OsAAN4 and OsGLR3.4. These results indicate that Cd transporters are in part responsible for Cd2+ entry into rice root, and provide a new perspective that the Ca channels OsAAN4 and OsGLR3.4 might play an important role in rice root Cd uptake.
Product Description
Supelco
2-Nitrophenyl octyl ether, Selectophore, ≥99.0%
Supelco
Cadmium ionophore I, Selectophore
Supelco
Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, Selectophore