MilliporeSigma
  • Surface Sizing Treated MWCNTs and Its Effect on the Wettability, Interfacial Interaction and Flexural Properties of MWCNT/Epoxy Nanocomposites.
Surface Sizing Treated MWCNTs and Its Effect on the Wettability, Interfacial Interaction and Flexural Properties of MWCNT/Epoxy Nanocomposites.
Nanomaterials (Basel, Switzerland) (2018-09-12)
Qingjie Zhang, Xinfu Zhao, Gang Sui, Xiaoping Yang
ABSTRACT
A surface-sizing technique was offered to take full advantage of multi-walled carbon nanotubes (MWCNTs) and epoxy resins. Two surface-sizing treated MWCNTs were obtained through a ball-milling treatment of amino-functionalized MWCNTs (MWCNT-NH₂) with n-butyl glycidylether (BuGE) and benzyl glycidylether (BeGE). These were referred to as MWCNT-BuGE and MWCNT-BeGE. The results indicated that the surface sizing effectively enhanced wettability, dispersibility of MWCNTs in the epoxy resin. These ameliorating effects, along with improved interfacial interaction between MWCNT-BeGE containing benzene rings and the epoxy matrix, which can offer a more efficient local load-transfer from matrix to MWCNTs, as observed by a higher G-band shift in Raman spectrum under bending loads than that of MWCNT-BuGE reinforced ones. Correspondingly, MWCNT-BeGE/epoxy nanocomposites exhibited increasing flexural strength and modulus of 22.9% and 37.8% respectively compared with the neat epoxy, and 7.3% and 7.7% respectively compared with MWCNT-BuGE/epoxy nanocomposites with the same MWCNT content.
Sigma-Aldrich
Benzyl glycidyl ether, 99%