MilliporeSigma
  • A baru almond-enriched diet reduces abdominal adiposity and improves high-density lipoprotein concentrations: a randomized, placebo-controlled trial.
A baru almond-enriched diet reduces abdominal adiposity and improves high-density lipoprotein concentrations: a randomized, placebo-controlled trial.
Nutrition (Burbank, Los Angeles County, Calif.) (2018-08-08)
Rávila Graziany Machado de Souza, Aline Corado Gomes, Inar Alves de Castro, João Felipe Mota
ABSTRACT
Nut consumption is associated with reduced risks of cardiovascular disease. Baru almonds have a high protein content and high quantities of mono- and polyunsaturated fatty acids, phenolic compounds, and antioxidants. This study aimed to evaluate the effects of a baru almond-enriched diet on body composition and markers of lipid metabolism in overweight and obese women. A randomized, placebo-controlled, 8-wk clinical trial of 46 overweight and obese women was conducted. Participants were randomly assigned to 1 of 2 normocaloric and isoenergetic diets: baru almond-enriched diet or baru almond-free diet. Both groups received dietary instructions. Body composition was assessed by anthropometry and dual-energy x-ray absorptiometry. Blood pressure, glucose levels, lipid profile, and plasma fatty acids, as well as apolipoproteins, angiopoietin-like-3, and cholesteryl ester transfer protein expression, were determined at the beginning and end of the study. The consumption of baru almonds reduced waist circumference (-2.45 cm; 95% confidence interval [CI], -3.90 to -0.23; P = 0.03), cholesteryl ester transfer protein expression (-0.23 mcg/mL; 95% CI, -1.24 to-0.08; P = 0.03), and increased high-density lipoprotein concentrations (+4.82 mg/dL; 95% CI, 0.03-8.88; P = 0.04) compared with baru almond-free diet. A baru almond-enriched diet for 8-wk reduced abdominal adiposity and improved high-density lipoprotein in overweight and obese women. This trial was registered at clinicaltrials.gov as RBR-2 wpryx.
Product Description
Supelco
C4 - C24 Even Carbon Saturated FAMEs, 1000 μg/mL each component in hexane, analytical standard