MilliporeSigma
  • Pseudo-Continuous Flow FTIR System for Glucose, Fructose and Sucrose Identification in Mid-IR Range.
Pseudo-Continuous Flow FTIR System for Glucose, Fructose and Sucrose Identification in Mid-IR Range.
Micromachines (2018-11-15)
Hamza Landari, Mourad Roudjane, Younès Messaddeq, Amine Miled
ABSTRACT
In this paper, we present a new FTIR-based microfluidic system for Glucose, Fructose and Sucrose detection. The proposed microfluidic system is based on a pseudo-continuous flow coupled to a microscope-FTIR instrument. The detection and characterization of sugar samples were performed by recording their absorption spectrum in the wavelength range 700⁻1000 cm - 1 of the Mid-IR region. The proposed pseudo-continuous flow system is designed to improve the uniformity of the sample distribution in the analyzed area versus conventional systems. The obtained results for different sugars concentrations, show a very low measurement error of 4.35% in the absorption peak intensity, which is ten times lower than the error obtained using the conventional measurements.
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)