MilliporeSigma
  • Characterization of a potent and highly unusual minimally enhancing antibody directed against dengue virus.
Characterization of a potent and highly unusual minimally enhancing antibody directed against dengue virus.
Nature immunology (2018-10-17)
Max Renner, Aleksandra Flanagan, Wanwisa Dejnirattisai, Chunya Puttikhunt, Watchara Kasinrerk, Piyada Supasa, Wiyada Wongwiwat, Kriangkrai Chawansuntati, Thaneeya Duangchinda, Alison Cowper, Claire M Midgley, Prida Malasit, Juha T Huiskonen, Juthathip Mongkolsapaya, Gavin R Screaton, Jonathan M Grimes
ABSTRACT
Dengue virus is a major pathogen, and severe infections can lead to life-threatening dengue hemorrhagic fever. Dengue virus exists as four serotypes, and dengue hemorrhagic fever is often associated with secondary heterologous infections. Antibody-dependent enhancement (ADE) may drive higher viral loads in these secondary infections and is purported to result from antibodies that recognize dengue virus but fail to fully neutralize it. Here we characterize two antibodies, 2C8 and 3H5, that bind to the envelope protein. Antibody 3H5 is highly unusual as it not only is potently neutralizing but also promotes little if any ADE, whereas antibody 2C8 has strong capacity to promote ADE. We show that 3H5 shows resilient binding in endosomal pH conditions and neutralizes at low occupancy. Immunocomplexes of 3H5 and dengue virus do not efficiently interact with Fcγ receptors, which we propose is due to the binding mode of 3H5 and constitutes the primary mechanism of how ADE is avoided.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution