MilliporeSigma
  • Highly selective asymmetric acetate aldol reactions of an N-acetyl thiazolidinethione reagent.
Highly selective asymmetric acetate aldol reactions of an N-acetyl thiazolidinethione reagent.
Organic letters (2004-01-03)
Yingchao Zhang, Andrew J Phillips, Tarek Sammakia
ABSTRACT
[reaction: see text] A highly diastereoselective acetate aldol reaction that uses a tert-leucine-derived thiazolidinethione auxiliary and dichlorophenylborane has been developed. The reaction proceeds in excellent yields and with high diastereoselectivities (drs range from 9.5:1 to >100:1).
Sigma-Aldrich
L-tert-Leucine, 99%