MilliporeSigma
  • Catalyst-free activation of kraft lignin in air using hydrogen sulfate ionic liquids.
Catalyst-free activation of kraft lignin in air using hydrogen sulfate ionic liquids.
International journal of biological macromolecules (2018-07-29)
Tadeusz J Szalaty, Łukasz Klapiszewski, Małgorzata Stanisz, Dariusz Moszyński, Andrzej Skrzypczak, Teofil Jesionowski
ABSTRACT
In this research we use ionic liquids in combination with mild process conditions to provide a selective increase in the content of carbonyl groups in the kraft lignin structure. Such modification can improve the properties of the pristine biopolymer. In this study, aromatic substituted ionic liquids were synthesized using [C4C1Im][HSO4] as a template structure. The substituents were intended to increase the affinity of the ionic liquid to the aromatic structure of kraft lignin, and to increase access to the oxidizing agent, which was atmospheric oxygen. [Benzyl(C2OC1)Im][HSO4] and [benzylC4Im][HSO4] activate the surface of the biopolymer more effectively than [C4C1Im][HSO4]. This was confirmed based on X-ray photoelectron spectroscopy, which showed the content of CO groups to be almost doubled compared with unmodified lignin. The analysis also revealed targeting of the hydroxyl groups of lignin to the carboxyl groups during activation in [C4C1Im][HSO4].
Sigma-Aldrich
1-Benzylimidazole, 99%
Sigma-Aldrich
Chloromethyl ethyl ether, 95%