MilliporeSigma
  • Toxicity of the biocide polycarbamate, used for aquaculture nets, to some marine fish species.
Toxicity of the biocide polycarbamate, used for aquaculture nets, to some marine fish species.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2018-09-12)
Kazuhiko Mochida, Katsutoshi Ito, Mana Ito, Takeshi Hano, Nobuyuki Ohkubo
ABSTRACT
We investigated toxic effects of the antifouling biocide polycarbamate (PC) on marine fish by conducting acute, early-life stage toxicity (ELS), and embryo toxicity tests. Mummichog (Fundulus heteroclitus) 96-h LC50 values for hatched larvae (body weight about 2.0 mg) and juveniles (660 ± 36 mg) were about 12 and 630 μg/L, respectively. The ELS test using mummichog embryos yielded a lowest-observed-effect concentration of 3.9 μg/L and a no-observed-effect concentration of 2.1 μg/L with growth as the most sensitive endpoint. The embryo toxicity test for spotted halibut (Verasper variegatus) revealed a 10-d EC50 of 8.1 μg/L with abnormality as an endpoint. During the ELS and embryo toxicity tests, morphological abnormalities (notochord undulation) were induced in the embryos. Biochemical and gene-expression analysis suggest that PC-induced morphological abnormalities involve disruption of lysyl oxidase-mediated collagen fiber organization, essential for notochord formation, and inhibition of gene expression related to notochord formation.
Sigma-Aldrich
Ampliflu Red, for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
2-Mercaptopyridine N-oxide, 99%