MilliporeSigma
  • Lumichrome inhibits osteoclastogenesis and bone resorption through suppressing RANKL-induced NFAT activation and calcium signaling.
Lumichrome inhibits osteoclastogenesis and bone resorption through suppressing RANKL-induced NFAT activation and calcium signaling.
Journal of cellular physiology (2018-06-16)
Chuan Liu, Zhen Cao, Wen Zhang, Jennifer Tickner, Heng Qiu, Chao Wang, Kai Chen, Ziyi Wang, Renxiang Tan, Shiwu Dong, Jiake Xu
ABSTRACT
The dynamic balance between bone resorption and bone formation is crucial to maintain bone mass. Osteoclasts are key cells that perform bone resorption while osteoblasts and osteocytes function in bone formation. Osteoporosis, a bone metabolism disease characterized by bone loss and degradation of bone microstructure, occurs when osteoclastic bone resorption outstrips osteoblastic bone synthesis. The interaction between receptor activator of nuclear factor κB ligand (RANKL) and RANK on the surface of bone marrow macrophages promotes osteoclast differentiation and activation. In this study, we found that lumichrome, a photodegradation product of riboflavin, inhibits RANKL-induced osteoclastogenesis and bone resorption as determined by tartrate-resistant acid phosphatase staining, immunofluorescence, reverse transcription-polymerase chain reaction, and western blot. Our results showed that lumichrome represses the expression of osteoclast marker genes, including cathepsin K (Ctsk) and Nfatc1. In addition, lumichrome suppressed RANKL-induced calcium oscillations, NFATc1, NF-κB, and MAPK signaling activation. Moreover, lumichrome promoted osteoblast differentiation at an early stage, as demonstrated by upregulated expression of osteoblast marker genes Alp, Runx2, and Col1a1. We also found that lumichrome reduces bone loss in ovariectomized mice by inhibiting osteoclastogenesis. In summary, our data suggest the potential of lumichrome as a therapeutic drug for osteolytic diseases.
Sigma-Aldrich
Lumichrome