- Overlap in the pharmacology of L-type Ca2+-channel blockers and 5-HT2 receptor antagonists in rat aorta.
Overlap in the pharmacology of L-type Ca2+-channel blockers and 5-HT2 receptor antagonists in rat aorta.
We have previously shown that elimination of buffer Ca2+ markedly reduced maximum 5-HT-induced contractions. We have now investigated the effect of L-type Ca2+-channel blockers and 5-HT2 receptor antagonists on 5-HT- and K+-induced contractions in rat aorta to explore the possibility of a relationship between blockade of L-type Ca2+ channels and 5-HT2 receptor antagonism. Sodium nitroprusside, felodipine, nifedipine, diltiazem, cinnarizine, verapamil, ritanserin, cyproheptadine, ketanserin and mianserin inhibited 5-HT-induced contractions of rat aorta with mean IC50 values (concentration (M) resulting in 50% inhibition) of 2.2 x 10(-11), 6.6 x 10(-11), 1.5 x 10(-9), 1.7 x 10(-9), 3.2 x 10(-7), 5.4 x 10(-7), 9.7 x 10(-10), 1.9 x 10(-8), 5.0 x 10(-7) and 6.4 x 10(-7), respectively. The same compounds antagonized K+-induced rat aortic contractions with the rank order of potency (mean IC50, M): felodipine (7.0 x 10(-11)) > nifedipine (4.8 x 10(-9)) > sodium nitroprusside (4.1 x 10(-8)) > verapamil (5.5 x 10(-8)) > cyproheptadine (6.2 x 10(-8)) > diltiazem (4.1 x 10(-7)) > cinnarizine (1.3 x 10(-6)) > ritanserin (1.8 x 10(-6)) > ketanserin (9.0 x 10(-6)) > mianserin (2.0 x 10(-5)). These data are indicative of a highly significant correlation (r=0.81, P=0.03) between potency against 5-HT-induced contraction and that against contractile response to K+ depolarization, and suggest overlap of the pharmacology of L-type Ca2+-channel blockers and 5-HT2 receptor antagonists in rat aorta.