MilliporeSigma
  • Suppression of the synaptic localization of a subset of proteins including APP partially ameliorates phenotypes of the Drosophila Alzheimer's disease model.
Suppression of the synaptic localization of a subset of proteins including APP partially ameliorates phenotypes of the Drosophila Alzheimer's disease model.
PloS one (2018-09-19)
Koto Furotani, Keisuke Kamimura, Takaaki Yajima, Minoru Nakayama, Rena Enomoto, Takuya Tamura, Hitoshi Okazawa, Masaki Sone
ABSTRACT
APP (amyloid precursor protein), the causative molecule of Alzheimer's disease, is synthesized in neuronal cell bodies and subsequently transported to synapses. We previously showed that the yata gene is required for the synaptic transport of the APP orthologue in Drosophila melanogaster. In this study, we examined the effect of a reduction in yata expression in the Drosophila Alzheimer's disease model, in which expression of human mutant APP was induced. The synaptic localization of APP and other synaptic proteins was differentially inhibited by yata knockdown and null mutation. Expression of APP resulted in abnormal synaptic morphology and the premature death of animals. These phenotypes were partially but significantly rescued by yata knockdown, whereas yata knockdown itself caused no abnormality. Moreover, we observed that synaptic transmission accuracy was impaired in our model, and this phenotype was improved by yata knockdown. Thus, our data suggested that the phenotypes caused by APP can be partially prevented by inhibition of the synaptic localization of a subset of synaptic proteins including APP.
Sigma-Aldrich
Anti-Peroxidase antibody produced in rabbit, fractionated antiserum, lyophilized powder