MilliporeSigma
  • A Single Adaptable Cochaperone-Scaffold Complex Delivers Nascent Iron-Sulfur Clusters to Mammalian Respiratory Chain Complexes I-III.
A Single Adaptable Cochaperone-Scaffold Complex Delivers Nascent Iron-Sulfur Clusters to Mammalian Respiratory Chain Complexes I-III.
Cell metabolism (2017-04-06)
Nunziata Maio, Ki Soon Kim, Anamika Singh, Tracey A Rouault
ABSTRACT
The iron-sulfur (Fe-S) cluster of the Rieske protein, UQCRFS1, is essential for Complex III (CIII) activity, though the mechanism for Fe-S cluster transfer has not previously been elucidated. Recent studies have shown that the co-chaperone HSC20, essential for Fe-S cluster biogenesis of SDHB, directly binds LYRM7, formerly described as a chaperone that stabilizes UQCRFS1 prior to its insertion into CIII. Here we report that a transient subcomplex involved in CIII assembly, composed of LYRM7 bound to UQCRFS1, interacts with components of an Fe-S transfer complex, consisting of HSC20, its cognate chaperone HSPA9, and the holo-scaffold ISCU. Binding of HSC20 to the LYR motif of LYRM7 in a pre-assembled UQCRFS1-LYRM7 intermediate in the mitochondrial matrix facilitates Fe-S cluster transfer to UQCRFS1. The five Fe-S cluster subunits of Complex I also interact with HSC20 to acquire their clusters, highlighting the crucial role of HSC20 in the assembly of the mitochondrial respiratory chain.
Sigma-Aldrich
Anti-Lipoic Acid Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-HSCB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-HSPA9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid