MilliporeSigma
  • Diurnal Oscillations in Liver Mass and Cell Size Accompany Ribosome Assembly Cycles.
Diurnal Oscillations in Liver Mass and Cell Size Accompany Ribosome Assembly Cycles.
Cell (2017-05-06)
Flore Sinturel, Alan Gerber, Daniel Mauvoisin, Jingkui Wang, David Gatfield, Jeremy J Stubblefield, Carla B Green, Frédéric Gachon, Ueli Schibler
ABSTRACT
The liver plays a pivotal role in metabolism and xenobiotic detoxification, processes that must be particularly efficient when animals are active and feed. A major question is how the liver adapts to these diurnal changes in physiology. Here, we show that, in mice, liver mass, hepatocyte size, and protein levels follow a daily rhythm, whose amplitude depends on both feeding-fasting and light-dark cycles. Correlative evidence suggests that the daily oscillation in global protein accumulation depends on a similar fluctuation in ribosome number. Whereas rRNA genes are transcribed at similar rates throughout the day, some newly synthesized rRNAs are polyadenylated and degraded in the nucleus in a robustly diurnal fashion with a phase opposite to that of ribosomal protein synthesis. Based on studies with cultured fibroblasts, we propose that rRNAs not packaged into complete ribosomal subunits are polyadenylated by the poly(A) polymerase PAPD5 and degraded by the nuclear exosome.
Sigma-Aldrich
Anti-RPS29 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-PAPD5 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Anti-U2AF65 antibody, Mouse monoclonal, clone MC3, purified from hybridoma cell culture