MilliporeSigma
  • Phosphonic acid-containing analogues of mycophenolic acid as inhibitors of IMPDH.
Phosphonic acid-containing analogues of mycophenolic acid as inhibitors of IMPDH.
Bioorganic & medicinal chemistry letters (2006-04-20)
William J Watkins, James M Chen, Aesop Cho, Lee Chong, Nicole Collins, Maria Fardis, Wei Huang, Magdeleine Hung, Thorsten Kirschberg, William A Lee, Xiaohong Liu, William Thomas, Jie Xu, Ameneh Zeynalzadegan, Jennifer Zhang
ABSTRACT
The design, synthesis, and IMPDH inhibitory activity of a series of phosphonic acid-containing analogues of mycophenolic acid are described.
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture