MilliporeSigma
  • Polyamine flux suppresses histone lysine demethylases and enhances ID1 expression in cancer stem cells.
Polyamine flux suppresses histone lysine demethylases and enhances ID1 expression in cancer stem cells.
Cell death discovery (2018-11-21)
Keisuke Tamari, Masamitsu Konno, Ayumu Asai, Jun Koseki, Kazuhiko Hayashi, Koichi Kawamoto, Noriyuki Murai, Senya Matsufuji, Fumiaki Isohashi, Taroh Satoh, Noriko Goto, Shinji Tanaka, Yuichiro Doki, Masaki Mori, Kazuhiko Ogawa, Hideshi Ishii
ABSTRACT
Cancer stem cells (CSCs) exhibit tumorigenic potential and can generate resistance to chemotherapy and radiotherapy. A labeled ornithine decarboxylase (ODC, a rate-limiting enzyme involved in polyamine [PA] biosynthesis) degradation motif (degron) system allows visualization of a fraction of CSC-like cells in heterogeneous tumor populations. A labeled ODC degradation motif system allowed visualization of a fraction of CSC-like cells in heterogeneous tumor populations. Using this system, analysis of polyamine flux indicated that polyamine metabolism is active in CSCs. The results showed that intracellular polyamines inhibited the activity of histone lysine 4 demethylase enzymes, including lysine-specific demethylase-1 (LSD1). Chromatin immunoprecipitation with Pol II antibody followed by massively parallel DNA sequencing, revealed the global enrichment of Pol II in transcription start sites in CSCs. Increase of polyamines within cells resulted in an enhancement of ID1 gene expression. The results of this study reveal details of metabolic pathways that drive epigenetic control of cancer cell stemness and determine effective therapeutic targets in CSCs.
Sigma-Aldrich
Anti-SMS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
Sigma-Aldrich
Spermidine, BioReagent, suitable for cell culture, ≥98%