MilliporeSigma
  • Inhibitors of lipid phosphatidate receptors: N-palmitoyl-serine and N-palmitoyl-tyrosine phosphoric acids.
Inhibitors of lipid phosphatidate receptors: N-palmitoyl-serine and N-palmitoyl-tyrosine phosphoric acids.
Journal of lipid research (1996-02-01)
R Bittman, B Swords, K Liliom, G Tigyi
ABSTRACT
An improved synthesis of two lipid phosphoric acids, N-palmitoyl-L-serine phosphoric acid (NP-Ser-PA) and N-palmitoyl-L-tyrosine phosphoric acid (NP-Tyr-PA), from the benzyl esters of L-serine and L-tyrosine is described. The sequence of N-acylation, followed by phosphitylation with N, N-diisopropyl dibenzyl phosphoramidite, oxidation to the corresponding phosphate triesters, and simultaneous debenzylation of the dibenzyl phosphate and benzyl carboxylic esters gave NP-Ser-PA and NP-Tyr-PA in high overall yields. NP-Ser-PA and NP-Tyr-PA and their D stereoisomers were potent reversible inhibitors of the lysophosphatidic acid receptors expressed in Xenopus oocytes, thus providing prototypic structures for the development of inhibitors of the lysophosphatidate family of phospholipid growth factors.
Product Description
Avanti
N-P Tyrosine PA, Avanti Research - A Croda Brand 800725P, powder
Avanti
N-P Serine PA, Avanti Research - A Croda Brand 800715P, powder