MilliporeSigma
  • Effects of Ceramide and Dihydroceramide Stereochemistry at C-3 on the Phase Behavior and Permeability of Skin Lipid Membranes.
Effects of Ceramide and Dihydroceramide Stereochemistry at C-3 on the Phase Behavior and Permeability of Skin Lipid Membranes.
Langmuir : the ACS journal of surfaces and colloids (2017-12-13)
Andrej Kováčik, Petra Pullmannová, Jaroslav Maixner, Kateřina Vávrová
ABSTRACT
Ceramides (Cer) are key components of the skin permeability barrier. Sphingosine-based CerNS and dihydrosphingosine-based CerNdS (dihydroCer) have two chiral centers; however, the importance of the correct stereochemistry in the skin barrier Cer is unknown. We investigated the role of the configuration at C-3 of CerNS and CerNdS in the organization and permeability of model skin lipid membranes. Unnatural l-threo-CerNS and l-threo-CerNdS with 24-C acyl chains were synthesized and, along with their natural d-erythro-isomers, incorporated into membranes composed of major stratum corneum lipids (Cer, free fatty acids, cholesterol, and cholesteryl sulfate). The membrane microstructure was investigated by X-ray powder diffraction and infrared spectroscopy, including deuterated free fatty acids. Inversion of the C-3 configuration in CerNS and CerNdS increased phase transition temperatures, had no significant effects on lamellar phases, but also decreased the proportion of orthorhombic packing and decreased lipid mixing in the model membranes. These changes in membrane organization resulted in membrane permeabilities that ranged from unchanged to 5-fold higher (depending on the permeability markers, namely, water loss, electrical impedance, flux of theophylline, and flux of indomethacin) compared to membranes with natural CerNS/NdS isomers. Thus, the physiological d-erythro stereochemistry of skin Cer and dihydroCer appears to be essential for their correct barrier function.
Product Description
Avanti
L-threo-sphingosine (d18:1), Avanti Research - A Croda Brand 860489P, powder
Avanti
Safingol, Avanti Research - A Croda Brand 860488P, powder