MilliporeSigma
  • Characterization of a dynamic metabolon producing the defense compound dhurrin in sorghum.
Characterization of a dynamic metabolon producing the defense compound dhurrin in sorghum.
Science (New York, N.Y.) (2016-11-20)
Tomas Laursen, Jonas Borch, Camilla Knudsen, Krutika Bavishi, Federico Torta, Helle Juel Martens, Daniele Silvestro, Nikos S Hatzakis, Markus R Wenk, Timothy R Dafforn, Carl Erik Olsen, Mohammed Saddik Motawia, Björn Hamberger, Birger Lindberg Møller, Jean-Etienne Bassard
ABSTRACT
Metabolic highways may be orchestrated by the assembly of sequential enzymes into protein complexes, or metabolons, to facilitate efficient channeling of intermediates and to prevent undesired metabolic cross-talk while maintaining metabolic flexibility. Here we report the isolation of the dynamic metabolon that catalyzes the formation of the cyanogenic glucoside dhurrin, a defense compound produced in sorghum plants. The metabolon was reconstituted in liposomes, which demonstrated the importance of membrane surface charge and the presence of the glucosyltransferase for metabolic channeling. We used in planta fluorescence lifetime imaging microscopy and fluorescence correlation spectroscopy to study functional and structural characteristics of the metabolon. Understanding the regulation of biosynthetic metabolons offers opportunities to optimize synthetic biology approaches for efficient production of high-value products in heterologous hosts.
Product Description
Avanti
14:0 PE, Avanti Research - A Croda Brand
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, powder
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
16:0 PA, Avanti Research - A Croda Brand
Avanti
C17 Ceramide (d18:1/17:0), Avanti Research - A Croda Brand
Avanti
14:0 PS, Avanti Research - A Croda Brand
Avanti
14:0 PG, Avanti Research - A Croda Brand