MilliporeSigma
  • Photo-Oxidation of Unilamellar Vesicles by a Lipophilic Pterin: Deciphering Biomembrane Photodamage.
Photo-Oxidation of Unilamellar Vesicles by a Lipophilic Pterin: Deciphering Biomembrane Photodamage.
Langmuir : the ACS journal of surfaces and colloids (2018-11-21)
Mariana Vignoni, Maria Noel Urrutia, Helena C Junqueira, Alexander Greer, Ana Reis, Mauricio S Baptista, Rosangela Itri, Andrés H Thomas
ABSTRACT
Pterins are natural products that can photosensitize the oxidation of DNA, proteins, and phospholipids. Recently, a new series of decyl-chain (i.e., lipophilic) pterins were synthesized and their photophysical properties were investigated. These decyl-pterins led to efficient intercalation in large unilamellar vesicles and produced, under UVA irradiation, singlet molecular oxygen, a highly oxidative species that react with polyunsaturated fatty acids (PUFAs) to form hydroperoxides. Here, we demonstrate that the association of 4-(decyloxy)pteridin-2-amine ( O-decyl-Ptr) to lipid membranes is key to its ability to trigger phospholipid oxidation in unilamellar vesicles of phosphatidylcholine rich in PUFAs used as model biomembranes. Our results show that O-decyl-Ptr is at least 1 order of magnitude more efficient photosensitizer of lipids than pterin (Ptr), the unsubstituted derivative of the pterin family, which is more hydrophilic and freely passes across lipid membranes. Lipid peroxidation photosensitized by O-decyl-Ptr was detected by the formation of conjugated dienes and oxidized lipids, such as hydroxy and hydroperoxide derivatives. These primary products undergo a rapid conversion into short-chain secondary products by cleavage of the fatty-acid chains, some of which are due to subsequent photosensitized reactions. As a consequence, a fast increase in membrane permeability is observed. Therefore, lipid oxidation induced by O-decyl-Ptr could promote cell photodamage due to the biomembrane integrity loss, which in turn may trigger cell death.
Product Description
Avanti
18:2 (Cis) PC (DLPC), 1,2-dilinoleoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
18:2 (Cis) PC (DLPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand