MilliporeSigma
  • An allosteric mechanism for activation of the kinase domain of epidermal growth factor receptor.
An allosteric mechanism for activation of the kinase domain of epidermal growth factor receptor.
Cell (2006-06-17)
Xuewu Zhang, Jodi Gureasko, Kui Shen, Philip A Cole, John Kuriyan
ABSTRACT
The mechanism by which the epidermal growth factor receptor (EGFR) is activated upon dimerization has eluded definition. We find that the EGFR kinase domain can be activated by increasing its local concentration or by mutating a leucine (L834R) in the activation loop, the phosphorylation of which is not required for activation. This suggests that the kinase domain is intrinsically autoinhibited, and an intermolecular interaction promotes its activation. Using further mutational analysis and crystallography we demonstrate that the autoinhibited conformation of the EGFR kinase domain resembles that of Src and cyclin-dependent kinases (CDKs). EGFR activation results from the formation of an asymmetric dimer in which the C-terminal lobe of one kinase domain plays a role analogous to that of cyclin in activated CDK/cyclin complexes. The CDK/cyclin-like complex formed by two kinase domains thus explains the activation of EGFR-family receptors by homo- or heterodimerization.
Product Description
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand