MilliporeSigma
  • Phosphatidylethanolamine and monoglucosyldiacylglycerol are interchangeable in supporting topogenesis and function of the polytopic membrane protein lactose permease.
Phosphatidylethanolamine and monoglucosyldiacylglycerol are interchangeable in supporting topogenesis and function of the polytopic membrane protein lactose permease.
The Journal of biological chemistry (2006-05-16)
Jun Xie, Mikhail Bogdanov, Philip Heacock, William Dowhan
ABSTRACT
To determine the specific role lipids play in membrane protein topogenesis in vivo, the orientation with respect to the membrane bilayer of Escherichia coli lactose permease (LacY) transmembrane (TM) domains and their flanking extramembrane domains was compared after assembly in native membranes and membranes with genetically modified lipid content using the substituted cysteine accessibility method for determining TM domain mapping. LacY assembled in the absence of the major membrane lipid phosphatidylethanolamine (PE) does not carry out uphill transport of substrate and displays an inverted orientation for the N-terminal six-TM domain helical bundle (Bogdanov, M., Heacock, P. N., and Dowhan, W. (2002) EMBO J. 21, 2107-2116). Strikingly, the replacement of PE in vivo by the foreign lipid monoglucosyldiacylglycerol (MGlcDAG), synthesized by the Acholeplasma laidlawii MGlcDAG synthase, restored uphill transport and supported the wild type TM topology of the N-terminal helical bundle of LacY. An interchangeable role in defining membrane protein TM domain orientation and supporting function is played by the two most abundant lipids, PE and MGlcDAG, in gram-negative and gram-positive bacteria, respectively. Therefore, these structurally diverse lipids endow the membrane with similar properties necessary for the proper organization of protein domains in LacY that are highly sensitive to lipids as topological determinants.