MilliporeSigma
Light-triggered enzymatic reactions in nested vesicle reactors.
Nature communications (2018-03-17)
James W Hindley, Yuval Elani, Catriona M McGilvery, Simak Ali, Charlotte L Bevan, Robert V Law, Oscar Ces
ABSTRACT
Cell-sized vesicles have tremendous potential both as miniaturised pL reaction vessels and in bottom-up synthetic biology as chassis for artificial cells. In both these areas the introduction of light-responsive modules affords increased functionality, for example, to initiate enzymatic reactions in the vesicle interior with spatiotemporal control. Here we report a system composed of nested vesicles where the inner compartments act as phototransducers, responding to ultraviolet irradiation through diacetylene polymerisation-induced pore formation to initiate enzymatic reactions. The controlled release and hydrolysis of a fluorogenic β-galactosidase substrate in the external compartment is demonstrated, where the rate of reaction can be modulated by varying ultraviolet exposure time. Such cell-like nested microreactor structures could be utilised in fields from biocatalysis through to drug delivery.
Product Description
Avanti
23:2 Diyne PC [DC(8,9)PC], Avanti Research - A Croda Brand 870016P, powder