MilliporeSigma
  • Lyme disease spirochaetes possess an aggrecan-binding protease with aggrecanase activity.
Lyme disease spirochaetes possess an aggrecan-binding protease with aggrecanase activity.
Molecular microbiology (2013-05-29)
Theresa M Russell, Barbara J B Johnson
ABSTRACT
Connective tissues are the most common area of colonization for the Lyme disease spirochaete Borrelia burgdorferi. Colonization is aided by the interaction between numerous bacterial adhesins with components of the extracellular matrix (ECM). Here we describe a novel interaction between B. burgdorferi and the major ECM proteoglycan found in joints, aggrecan. Using affinity chromatography and mass spectrometry we identify two borrelial aggrecan-binding proteins: the known ECM ligand Bgp (BB0588) and an uncharacterized protease BbHtrA (BB0104). Proteinase K studies demonstrate that BbHtrA is surface exposed. Immunoblots using sera from patients with both early and late Lyme disease establish that BbHtrA is expressed during human disease, immunogenic, and conserved in the three major Lyme disease spirochaete species. Consequences of the interaction between aggrecan and BbHtrA were examined by proteolysis assays. BbHtrA cleaves aggrecan at a site known to destroy aggrecan function and which has been previously observed in the synovial fluid of patients with Lyme arthritis. These data demonstrate that B. burgdorferi possess aggrecan-binding proteins which may provide the organism with additional capability to colonize connective tissues. Moreover, our studies provide the first evidence that B. burgdorferi possess proteolytic activity which may contribute to the pathogenesis of Lyme arthritis.
Sigma-Aldrich
Anti-HTRA4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-HTRA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution